Learn More
Sino Biological Anti-Integrin beta 1 Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10587MM05A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Integrin beta 1/ITGB1 (rh Integrin beta 1/ITGB1; Catalog#10587-H08H1; CAA30790.1; Met1-Asp728) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-1353