Learn More
Sino Biological Anti-Integrin beta 1 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10587MM06P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ITGB1 / Integrin beta-1 / CD29 (rh ITGB1 / Integrin beta-1 / CD29; Catalog#10587-H08H1; CAA30790.1; Met1-Asp728) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-2202