Learn More
ACROBiosystems ACROBiosystems Human JAB1 / COPS5 Protein, Fc Tag
Catalog No. 502107120
Encompass_Preferred
Recombinant Protein;100UG;Human JAB1 Protein, Fc Tag (JA1-H5553) is expressed from Baculovirus-Insect cells. It contains AA Ala 2 - Ser 334 (Accession # Q92905-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 63.9 kDa.
Catalog No. 50-210-7120 Supplier ACROBiosystems Supplier No. JA1H5553100UG
Recombinant Protein;100UG;Human JAB1 Protein, Fc Tag (JA1-H5553) is expressed from Baculovirus-Insect cells. It contains AA Ala 2 - Ser 334 (Accession # Q92905-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 63.9 kDa.