Learn More
Sino Biological Anti-JADE1/PHF17 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101515T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human JADE1/PHF17, and purified by antigen affinity chromatography.
Catalog No. 50-240-8648