Learn More
Sino Biological Anti-Jagged 1/JAG1 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 11648MM03P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human JAG1 / Jagged 1 (rh JAG1 / Jagged 1; Catalog#11648-H08H; NP_000205.1; Met1-Ser1046) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-3238