Learn More
Invitrogen™ Human Jagged 1 ELISA Kit
Description
Human Jagged 1 quantitates human Jagged 1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Jagged 1.
Specifications
Specifications
| Accession Number | ABE2; AGS; AHD; AWS; CD339; Gsfabe2; HJ1; Htu; JAG1; jagged 1; jagged canonical Notch ligand 1; Jagged1; JAGL1; Ozz; protein jagged-1; Ser-1; Serrate-1 |
| Assay Range | 1.6 to 400 ng/mL |
| Assay Sensitivity | 1.6 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 182 |
| Gene Symbol | JAG1 |
| Interassay CV | <12% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.