Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human JAKMIP1 (aa 332-414) Control Fragment Recombinant Protein

Catalog No. RP96694
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96694 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96694 Supplier Invitrogen™ Supplier No. RP96694
  • $283.00
    $212.25 / Each of 1

    Save $70.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60756. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

JAKMIP1, also known as Marlin-1, is a highly-conserved vertebrate-specific microtubule-associated protein that plays a role in JAK1 signaling and regulate microtubule cytoskeleton rearrangements. JAKMIP1 is expressed predominantly in neuronal and lymphoid cells and colocalizes with microtubules. Multiple isoforms of JAKMIP1 are known to exist: isoforms 1 and 5 are also expressed in liver, lung and skeletal muscle, while isoforms 2-4 are specifically expressed in brain and retina. JAKMIP1 participates in polarized secretion in lymphocytes and it has been suggested to be a novel effector memory gene restraining T cell-mediated cytotoxicity. Dysregulation of JAKMIP1 gene expression is associated with genetic conditions linked to autism in humans.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96N16
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 152789
Name Human JAKMIP1 (aa 332-414) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 5830437M04Rik; C330021K24Rik; FLJ31564; GABA-B receptor-binding protein; Gababrbp; gamma-aminobutyric acid (GABA-B) receptor binding protein; Jak and microtubule interacting protein 1; Jakmip1; JAMIP1; janus kinase and microtubule interacting protein 1; janus kinase and microtubule-interacting protein 1; MARLIN1; Marlin-1; Multiple alpha helices and RNA-linker protein 1; multiple alpha-helices and RNA-linker protein 1; multiple coiled-coil GABABR1-binding protein
Common Name JAKMIP1
Gene Symbol JAKMIP1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LVEKNKRMNKKNEDLLQSIQRMEEKIKNLTRENVEMKEKLSAQASLKRHTSLNDLSLTRDEQEIEFLRLQVLEQQHVIDDLSL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.