Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human JAKMIP1 Synthetic Peptide

Catalog No. PEP1537 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1537 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1537 Supplier Invitrogen™ Supplier No. PEP1537
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 16 amino acids near the center of human JAKMIP1. PEP-1537 can be used as a blocking peptide with polyclonal antibody PA5-34495.

JAKMIP1, also known as Marlin-1, is a highly-conserved vertebrate-specific microtubule-associated protein that plays a role in JAK1 signaling and regulate microtubule cytoskeleton rearrangements. JAKMIP1 is expressed predominantly in neuronal and lymphoid cells and colocalizes with microtubules. Multiple isoforms of JAKMIP1 are known to exist: isoforms 1 and 5 are also expressed in liver, lung and skeletal muscle, while isoforms 2-4 are specifically expressed in brain and retina. JAKMIP1 participates in polarized secretion in lymphocytes and it has been suggested to be a novel effector memory gene restraining T cell-mediated cytotoxicity. Dysregulation of JAKMIP1 gene expression is associated with genetic conditions linked to autism in humans.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 152789
Species Human
Purification Method purified
Accession Number Q96N16
Common Name JAKMIP1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 5830437M04Rik; C330021K24Rik; FLJ31564; GABA-B receptor-binding protein; Gababrbp; gamma-aminobutyric acid (GABA-B) receptor binding protein; Jak and microtubule interacting protein 1; Jakmip1; JAMIP1; janus kinase and microtubule interacting protein 1; janus kinase and microtubule-interacting protein 1; MARLIN1; Marlin-1; Multiple alpha helices and RNA-linker protein 1; multiple alpha-helices and RNA-linker protein 1; multiple coiled-coil GABABR1-binding protein
Gene Symbol JAKMIP1
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.