Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human JAKMIP2 (aa 393-457) Control Fragment Recombinant Protein

Catalog No. RP100239
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100239 Supplier Invitrogen™ Supplier No. RP100239
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61374. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

JAKMIP2 (janus kinase and microtubule-interacting protein 2), also known as NECC1 (neuroendocrine long coiled-coil protein 1), CTCL tumor antigen HD-CL-04, JAMIP2 or KIAA0555, is a 810 amino acid protein belonging to the JAKMIP family. Localizing to the Golgi apparatus, JAKMIP2 is high expressed in brain, with moderate levels of expression found in thymus, spleen and lung. Existing as three alternatively spliced isoforms, the gene encoding JAKMIP2 maps to human chromosome 5q32 and mouse chromosome 18 B3. Chromosome 5 contains 181 million base pairs and comprises nearly 6% of the human genome. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96AA8
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 9832
Name Human JAKMIP2 (aa 393-457) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 6430702L21Rik; AI850334; CTCL tumor antigen HD-CL-04; D930046L20Rik; Jak and microtubule interacting protein 2; JAKMIP2; JAMIP2; janus kinase and microtubule interacting protein 2; janus kinase and microtubule-interacting protein 2; KIAA0555; NECC1; Neuroendocrine long coiled-coil protein 1; RGD1559742
Common Name JAKMIP2
Gene Symbol Jakmip2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KLQVIEQQNIIDELTRDREKLIRRRKHRRSSKPIKRPVLDPFIGYDEDSMDSETSSMASFRTDRT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.