Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human JAKMIP2 Synthetic Peptide

Catalog No. PEP1538
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1538 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1538 Supplier Invitrogen™ Supplier No. PEP1538
Only null left
Add to Cart
Add to Cart

Synthetic Peptide

JAKMIP2 Synthetic Peptide, PEP-1538, from Invitrogen.

JAKMIP2 (janus kinase and microtubule-interacting protein 2), also known as NECC1 (neuroendocrine long coiled-coil protein 1), CTCL tumor antigen HD-CL-04, JAMIP2 or KIAA0555, is a 810 amino acid protein belonging to the JAKMIP family. Localizing to the Golgi apparatus, JAKMIP2 is high expressed in brain, with moderate levels of expression found in thymus, spleen and lung. Existing as three alternatively spliced isoforms, the gene encoding JAKMIP2 maps to human chromosome 5q32 and mouse chromosome 18 B3. Chromosome 5 contains 181 million base pairs and comprises nearly 6% of the human genome. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9832
Species Human
Purification Method purified
Accession Number Q96AA8
Common Name Human JAKMIP2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol JAKMIP2
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.