Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human JAM-A/CD321/F11R ELISA Kit

Catalog No. EH283RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH283RBX5 5 x 96 Tests
EH283RBX10 10 x 96 Tests
EH283RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH283RBX5 Supplier Invitrogen™ Supplier No. EH283RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human JAM-A/CD321/F11R quantitates human JAM-A/CD321/F11R in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human JAM-A/CD321/F11R.

CD321 (F11R, JAM-1, JAM-A) is a cell-surface adhesion molecule belonging to the Ig superfamily of receptors. CD321 is a 32-41 kD glycoprotein, possessing two extracellular Ig-like domains, a transmembrane region, and a short cytosolic tail. The protein assembles at the membrane as a homodimer, and can associate laterally with other membrane proteins including integrins. CD321 is expressed on a variety of cell types, including leukocytes, platelets, erythrocytes, stem cells, epithelial and endothelial cells, and plays a role in mediating cell-cell contact. CD321 can be found in close proximity to several components of epithelial tight junctions, including ZO-1 and occludin. CD321 is involved in the maintenance of epithelial and endothelial barriers, hematopoiesis, angiogenesis, and tumor proliferation, invasion, and metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y624
Assay Range 13-10000 pg/mL
Assay Sensitivity 13 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 9130004G24; AA638916; BV11 antigen; CD321; ESTM33; F11 R; F11 receptor; F11 receptor/F11R; F11R; JAM; JAM1; JAM-1; JAMA; JAM-A; Jcam; Jcam1; junction cell adhesion molecule A; junction cell adhesion molecule1; junctional adhesion molecule 1; Junctional adhesion molecule A; KAT; Ly106; PAM-1; Platelet adhesion molecule 1; platelet F11 receptor; UNQ264/PRO3019130004G24; AA638916; BV11 antigen; CD321; ESTM33; F11 R; F11 receptor; F11 receptor/F11R; F11R; JAM; JAM1; JAM-1; JAMA; JAM-A; Jcam; Jcam1; junction cell adhesion molecule A; junction cell adhesion molecule1; junctional adhesion molecule 1; Junctional adhesion molecule A; KAT; Ly106; PAM-1; Platelet adhesion molecule 1; platelet F11 receptor; UNQ264/PRO301;
Gene ID (Entrez) 50848
Gene Symbol F11r
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target JAM-A (CD321)
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.