Learn More
Sino Biological Human JAM-A/F11 Receptor Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785278
Encompass_Preferred
The Human Junctional Adhesion Molecule A / JAM-A ELISA Pair Set is for the quantitative determination of Human Junctional Adhesion Molecule A / JAM-A.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human Junctional Adhesion Molecule A / JAM-A coated on a 96-well plate. Standards and samples are added to the wells, and any Human Junctional Adhesion Molecule A / JAM-A present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human Junctional Adhesion Molecule A / JAM-A monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human Junctional Adhesion Molecule A / JAM-A present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read
Catalog No. 50-007-85278 Supplier Sino Biological Supplier No. SEKA101985
The Human Junctional Adhesion Molecule A / JAM-A ELISA Pair Set is for the quantitative determination of Human Junctional Adhesion Molecule A / JAM-A.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human Junctional Adhesion Molecule A / JAM-A coated on a 96-well plate. Standards and samples are added to the wells, and any Human Junctional Adhesion Molecule A / JAM-A present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human Junctional Adhesion Molecule A / JAM-A monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human Junctional Adhesion Molecule A / JAM-A present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read