Learn More
Raybiotech Inc Human JAM-C (CD323) ELISA Kit
Supplier:  Raybiotech Inc ELHJAMC1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.12 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Human Junctional Adhesion Molecule C (JAM3 / CD323)
Catalog No. 50-194-9475