Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human JAM-C ELISA Kit

Catalog No. EH285RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH285RBX5 5 x 96 Tests
EH285RB 96 Tests
EH285RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH285RBX5 Supplier Invitrogen™ Supplier No. EH285RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human JAM-C quantitates human JAM-C in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human JAM-C.

JAM3 is involved in tight junctions and represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. The protein encoded by this immunoglobulin superfamily gene member is localized in the tight junctions between high endothelial cells. Unlike other proteins in this family, the this protein is unable to adhere to leukocyte cell lines and only forms weak homotypic interactions. The encoded protein is a member of the junctional adhesion molecule protein family and acts as a receptor for another member of this family.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BX67
Assay Range 0.12-30 ng/mL
Assay Sensitivity 0.12 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 1110002N23Rik; JAM-2; JAM3; JAM-3; JAMC; JAM-C; Jcam3; junction adhesion molecule 3; junction cell adhesion molecule 3; junction cell adhesion molecule C; junctional adhesion molecule 3; Junctional adhesion molecule C; sJAM-C; Soluble form of JAM-C; UNQ859/PRO18681110002N23Rik; JAM-2; JAM3; JAM-3; JAMC; JAM-C; Jcam3; junction adhesion molecule 3; junction cell adhesion molecule 3; junction cell adhesion molecule C; junctional adhesion molecule 3; Junctional adhesion molecule C; sJAM-C; Soluble form of JAM-C; UNQ859/PRO1868;
Gene ID (Entrez) 83700
Gene Symbol JAM3
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target JAM3
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.