Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human JARID2 (aa 967-1050) Control Fragment Recombinant Protein

Catalog No. RP107294
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP107294 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP107294 Supplier Invitrogen™ Supplier No. RP107294
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Jumonji (JARID2) is a nuclear protein that participates in the negative regulation of cell growth. It is required for neural tube formation and is essential for normal heart development and function. Also, it is a part of a group of proteins characterized by a novel structural motif, the JmjC domain, which is implicated in chromatin regulation through histone demethylation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92833
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 3720
Name Human JARID2 (aa 967-1050) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias JARD2 antibody; Jarid2; Jmj; jumonji; jumonji and AT-rich interaction domain containing 2; jumonji homolog; jumonji, AT rich interactive domain 2; jumonji, AT rich interactive domain 2 protein; jumonji/ARID domain-containing protein 2; jumonji-like protein; LOW QUALITY PROTEIN: protein Jumonji; protein Jumonji
Common Name JARID2
Gene Symbol JARID2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LLQANGTPGLQMLESNVMISPEVLCKEGIKVHRTVQQSGQFVVCFPGSFVSKVCCGYSVSETVHFATTQWTSMGFETAKEMKRR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.