Learn More
Sino Biological Anti-KIAA1462 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 204715T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human KIAA1462 / JCAD fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-2151