Learn More
Biomatik Corporation Human K-Acetyltransferase 2B (KAT2B) ELISA Kit
Supplier:  Biomatik Corporation EKU12052
Encompass_Preferred
Human K-Acetyltransferase 2B (KAT2B) ELISA Kit for detecting KAT2B in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q92831. Gene ID: 8850. Target Synonyms: PCAF; P/CAF; GCN5; GCN5L; P300/CBP-Associated Factor; Histone acetyltransferase PCAF; Lysine acetyltransferase 2B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6109