Learn More
Biomatik Corporation Human Kallikrein 1 (KLK1) ELISA Kit
Supplier:  Biomatik Corporation EKU05428
Encompass_Preferred
Human Kallikrein 1 (KLK1) ELISA Kit for detecting KLK1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 74.07-6,000pg/ml. Sensitivity: 27.34pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P06870. Gene ID: 3816. Target Synonyms: KLKR; Klk6; HK1; Kallikrein 1,Renal/Pancreas/Salivary; Kidney/pancreas/salivary gland kallikrein; Tissue kallikrein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5043