Learn More
Enzyme Research Laboratories Inc HUMAN KALLIKREIN 1 MG VIAL
Catalog No. NC9018630
Encompass
Activation of Prekallikrein with Factor alpha -XIIa produces the enzymatically active Kallikrein. Kallikrein is a serine protease which consists of a heavy chain (Mr 52kD) and light chains (Mr either 36 or 33 kD) linked by disulfide bridges.
Catalog No. NC9018630 Supplier Enzyme Research Laboratories Inc Supplier No. HPKA 1303
Activation of Prekallikrein with Factor alpha -XIIa produces the enzymatically active Kallikrein. Kallikrein is a serine protease which consists of a heavy chain (Mr 52kD) and light chains (Mr either 36 or 33 kD) linked by disulfide bridges.