Learn More
Biomatik Corporation Human Kallikrein 14 (KLK14) ELISA Kit
Supplier:  Biomatik Corporation EKU05436
Encompass_Preferred
Human Kallikrein 14 (KLK14) ELISA Kit for detecting KLK14 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 123.46-10000pg/ml. Sensitivity: 43.2pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9P0G3. Gene ID: 43847. Target Synonyms: KLK-L6; Kallikrein-like protein 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8906