Learn More
Biomatik Corporation Human Kallikrein 4 (KLK4) ELISA Kit
Supplier:  Biomatik Corporation EKU11891
Encompass_Preferred
Human Kallikrein 4 (KLK4) ELISA Kit for detecting KLK4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y5K2. Gene ID: 9622. Target Synonyms: ARM1; EMSP1; KLK-L1; PRSS17; PSTS; Enamel Matrix,Prostate; Kallikrein-Related Peptidase 4; Enamel matrix serine proteinase 1; Serine protease 17; Kallikrein-like protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4561