Learn More
Biomatik Corporation Human Kallikrein 6 (KLK6) ELISA Kit
Supplier:  Biomatik Corporation EKU10149
Encompass_Preferred
Human Kallikrein 6 (KLK6) ELISA Kit for detecting KLK6 in serum, plasma, tissue homogenates, cerebrospinal fluid, breast milk, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q92876. Gene ID: 5653. Target Synonyms: ZYME; Bssp; Klk7; Neurosin,Zyme; PRSS18; PRSS9; SP59; HK6; Protease M; Serine protease 18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6366