Learn More
Biomatik Corporation Human Kallikrein 8 (KLK8) ELISA Kit
Supplier:  Biomatik Corporation EKU11677
Encompass_Preferred
Human Kallikrein 8 (KLK8) ELISA Kit for detecting KLK8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60259. Gene ID: 11202. Target Synonyms: NP; HNP; NRPN; PRSS19; TADG14; BSP1; Neuropsin; Ovasin; Brain Serine Protease 1; Tumor Associated Differentially Expressed Gene 14; Serine protease 19. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5365