Learn More
Biomatik Corporation Human Kallikrein 9 (KLK9) ELISA Kit
Supplier:  Biomatik Corporation EKU05453
Encompass_Preferred
Human Kallikrein 9 (KLK9) ELISA Kit for detecting KLK9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UKQ9. Gene ID: 284366. Target Synonyms: KLK-L3; Kallikrein-Related Peptidase 9; Kallikrein-like protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7716