Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human KANK3 Synthetic Peptide

Catalog No. PEP1583
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1583 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1583 Supplier Invitrogen™ Supplier No. PEP1583
Add to Cart
Add to Cart

Synthetic Peptide

KANK3 Synthetic Peptide, PEP-1583, from Invitrogen.

Ankyrins are membrane adaptor molecules that play important roles in the control of cytoskeleton formation by regulating actin polymerization. Like other members of the KANK family, KANK3 (KN motif and ankyrin repeat domain-containing protein 3), is thought to play a role in the formation of actin stress fibers. In zebrafish, the homolog of KANK3 interacts with the adaptor protein Numb, a protein implicated in multiple basic cellular processes, and is essential for epidermal integrity and neurulation, suggesting that KANK3 may play a similar role in higher organisms.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 256949
Species Human
Purification Method purified
Accession Number Q6NY19
Common Name Human KANK3
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol KANK3
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.