Learn More
Abcam Human Kappa ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1577091X96TESTS
Encompass_Preferred
Human Kappa ELISA Kit ab157709 is used to quantify Human Kappa with a sensitivity of 43 pg/ml. Sandwich ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1577312