Learn More
Biomatik Corporation Human Kappa Immunoglobulin Light Chain, Κ-Iglc Elisa Kit
Supplier:  Biomatik Corporation EKC31268
Encompass_Preferred
Human Kappa Immunoglobulin Light Chain, Κ-Iglc Elisa Kit for detecting Kappa Immunoglobulin Light Chain, Κ-Iglc in serum, plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 0.078-20ug/ml. Sensitivity: 0.046ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Target Synonyms: kappa immunoglobulin light chain; kappa-IgLC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9194