Learn More
ABclonal Technology Human Kappa Light Chain
SDP
Catalog No. 5000006301
Encompass_Preferred
Immunoglobulins are glycoproteins making up 20% of serum proteins playing a key role in immunity by reacting with antigens They consist of two identical heavy chains and two light chains each with constant and variable regions The constant regions are similar across all immunoglobulins while the variable regions have high sequence variability The heavy chain amino acid sequence defines the immunoglobulin class with five types IgG IgA IgM IgD and IgE IgG has four subclasses and IgA has two In serum IgA and IgG are monomers while IgM is a pentamer and IgA can form polymers The kappa light chain is used for diagnosing leukemias plasmacytomas and some non-Hodgkin s lymphomas It contains one immunoglobulin-like domain
Catalog No. 50-000-06301 Supplier ABclonal Technology Supplier No. A2368920UL
Immunoglobulins are glycoproteins making up 20% of serum proteins playing a key role in immunity by reacting with antigens They consist of two identical heavy chains and two light chains each with constant and variable regions The constant regions are similar across all immunoglobulins while the variable regions have high sequence variability The heavy chain amino acid sequence defines the immunoglobulin class with five types IgG IgA IgM IgD and IgE IgG has four subclasses and IgA has two In serum IgA and IgG are monomers while IgM is a pentamer and IgA can form polymers The kappa light chain is used for diagnosing leukemias plasmacytomas and some non-Hodgkin s lymphomas It contains one immunoglobulin-like domain