Learn More
Sino Biological Anti-LysRS/Lysyl tRNA synthetase Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 105296T42100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human LysRS/Lysyl tRNA synthetase/KARS, and purified by antigen affinity chromatography.
Catalog No. 50-241-1879