Learn More
Sino Biological Recombinant Anti-CSEN Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 14683R038100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CSEN / calsenilin / KCNIP3 (rh CSEN / calsenilin / KCNIP3; Catalog#14683-H07E; Q9Y2W7-1; Met1-Ile256).
Catalog No. 50-240-8151