Learn More
BIOHIPPO Human KCNJ10 Microsample
SDP
Catalog No. 5000314357
Encompass
Human KCNJ10 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of KCNJ10 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 6 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human KCNJ10 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human KCNJ10 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human KCNJ10 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14357 Supplier BIOHIPPO Supplier No. BHE15210827
Human KCNJ10 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of KCNJ10 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 6 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human KCNJ10 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human KCNJ10 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human KCNJ10 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color