Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human KCNMA1 (aa 887-902) Synthetic Peptide

Catalog No. PEP122 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP122 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP122 Supplier Invitrogen™ Supplier No. PEP122
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

PEP-122 is a 16 amino acid synthetic peptide whose sequences are derived from amino acids 887-902 of human Maxi K+ alpha protein. This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human NFAT3, catalog # PA1-021. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (~70-fold) for competitive inhibition of antibody-protein binding reactions. Reconstitute with 0.1 mL of distilled water.

Potassium channels are a group of ubiquitously expressed proteins that serve numerous functions in excitable and non-excitable cells. One class of integral membrane potassium channels is the large conductance, calcium-activated potassium channel (Maxi K+). Maxi K+ differs from most other potassium channels in that its activation is controlled by both increases in intracellular calcium and by membrane depolarization. Maxi K+ dual activation is possible because of its structure. The core of the channel, which is similar to other potassium channels, is a Maxi K+ alpha homotetramer that contains both a voltage sensor and an intracellular calcium binding domain. In vascular smooth muscle, an auxiliary beta-subunit is found in a 1:1 stoichiometry. The beta-subunit exhibits its effect on the Maxi K+ channel by effectively decreasing by 5- to 10- fold the concentration of calcium required to keep the pore open.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 3778
Species Human
Purification Method purified
Accession Number Q12791
Common Name KCNMA1
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation Peptide with no preservative
For Use With (Application) Control
Gene Alias 5730414M22Rik; bA205K10.1; big potassium channel alpha subunit; BK channel; BK channel alpha subunit; BKCa; BKCA alpha; BKCA alpha subunit; BKTM; calcium-activated potassium channel alpha subunit; calcium-activated potassium channel subunit alpha-1; calcium-activated potassium channel subunit alpha-1; uncharacterized protein; calcium-activated potassium channel, subfamily M subunit alpha-1; hSlo; K(VCA)alpha; KCa1.1; Kcnma; Kcnma1; KCNMA1b; KCNMA1c; Maxi K channel; maxiK; maxi-K channel HSLO; mSlo; mSlo1; potassium calcium-activated channel subfamily M alpha 1; potassium channel, calcium activated large conductance subfamily M alpha, member 1; potassium large conductance calcium-activated channel, subfamily M, alpha member 1; SAKCA; SLO; slo homolog; slo1; Slo-alpha; Slowpoke homolog; stretch-activated Kca channel
Gene Symbol Kcnma1
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Sequence Derived from Amino Acids 887-902 of Human Maxi K+ a Protein
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.