Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human KDM4E (aa 371-441) Control Fragment Recombinant Protein

Catalog No. RP100859
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100859 Supplier Invitrogen™ Supplier No. RP100859
  • $283.00
    $212.25 / Each of 1

    Save $70.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (32%), Rat (32%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this intronless gene is a member of a large family of histone lysine demethylases, which use oxygen and 2-oxoglutarate to demethylate di- and trimethylated lys9 of histone H3. Derepression of genes by demethylases is sometimes involved in viral infection or carcinogenesis, so inhibitors of these enzymes are desired.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number B2RXH2
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 390245
Name Human KDM4E (aa 371-441) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias JMJD2E; KDM4DL; Kdm4dl1; KDM4D-like protein; KDM4E; lysine (K)-specific demethylase 4D-like 1; lysine (K)-specific demethylase 4E; lysine demethylase 4E; lysine-specific demethylase 4D-like; Lysine-specific demethylase 4E
Common Name KDM4E
Gene Symbol KDM4E
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LRLLPNLTAQCPTQPVSSGHCYNPKGCGTDAVPGSAFQSSAYHTQTQSLTLGMSARVLLPSTGSWGSGRGR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.