Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human KDM5B (aa 1401-1481) Control Fragment Recombinant Protein

Catalog No. RP94108
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP94108 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP94108 Supplier Invitrogen™ Supplier No. RP94108
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55535. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

KDM5B is a histone demethylase that demethylates 'Lys-4' of histone H3, thereby playing a central role in histone code. It does not demethylate histone H3 'Lys-9' or H3 'Lys-27'. KDM5B demethylates trimethylated, dimethylated and monomethylated H3 'Lys-4'. It acts as a transcriptional corepressor for FOXG1B and PAX9. It favors the proliferation of breast cancer cells by repressing tumor suppressor genes such as BRCA1 and HOXA5. In contrast, KDM5B may act as a tumor suppressor for melanoma.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UGL1
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 10765
Name Human KDM5B (aa 1401-1481) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2010009J12Rik; 2210016I17Rik; AW556288; cancer/testis antigen 31; CT31; D1Ertd202e; Histone demethylase JARID1B; Jarid1b; jumonji, AT rich interactive domain 1B; jumonji, AT rich interactive domain 1B (Rbp2 like); Jumonji, AT rich interactive domain 1B (RBP2-like); jumonji/ARID domain-containing protein 1B; KDM5B; Kiaa4034; lysine (K)-specific demethylase 5B; lysine demethylase 5B; lysine-specific demethylase 5B; mKIAA4034; PLU1; PLU-1; PPP1R98; protein phosphatase 1, regulatory subunit 98; PUT1; putative DNA/chromatin binding motif; putative DNA/chromatin binding motif 1; Rb-Bp2; RBBP2H1; RBBP2H1A; RBP2-H1; Retinoblastoma-binding protein 2 homolog 1; retinoblastoma-binding protein 2, homolog 1A; RGD1565602
Common Name KDM5B
Gene Symbol KDM5B
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DGINSLERKLKRRLEREGLSSERWERVKKMRTPKKKKIKLSHPKDMNNFKLERERSYELVRSAETHSLPSDTSYSEQEDSE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.