Learn More
Sino Biological Recombinant Anti-VEGFR2/KDR Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10012R002100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human VEGFR2 / Flk-1 / CD309 / KDR (rh VEGFR2 / Flk-1 / CD309 / KDR; Catalog#10012-H08H; NP_002244.1; Met1-Glu764).
Catalog No. 50-241-0566