Learn More
Sino Biological Recombinant Human VEGFR2/KDR Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10012H08HB100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human VEGFR2 (NP_002244.1) (Met 1-Glu 764) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-5533