Learn More
Sino Biological Recombinant Human VEGFR2/KDR Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10012H02H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human VEGFR2 (NP_002244.1) (Met 1-Glu 764) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-5535