Learn More
Sino Biological Recombinant Human VEGFR2/KDR Protein (ECD, His & AVI Tag), Biotinylated
Supplier:  Sino Biological 10012H27HB20
Encompass_Preferred
A DNA sequence encoding the human KDR (NP_002244.1) (Met1-Glu764) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-5531