Learn More
Biomatik Corporation Human Keratin 6C (KRT6C) ELISA Kit
Supplier:  Biomatik Corporation EKU11028
Encompass_Preferred
Human Keratin 6C (KRT6C) ELISA Kit for detecting KRT6C in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48668. Gene ID: 286887. Target Synonyms: KRT6E; Keratin 6E; Keratin, type II cytoskeletal 6C; Cytokeratin-6C; Type-II keratin Kb12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8244