Learn More
Bioworld HUMAN KERATINOCYE GROWTH FA
Catalog No. 50253494
Encompass_Preferred
HUMAN KERATINOCYE GROWTH FACTOR (RECOMBINANT): Recombinant Human Keratinocyte Growth Factor (KGF) Source: E.Coli Background: KGF is a potent specific mitogen for many epithelial cells but not for fibroblasts and endothelial cells. KG...
Catalog No. 50-253-494 Supplier Bioworld Supplier No. 220603971
HUMAN KERATINOCYE GROWTH FACTOR (RECOMBINANT): Recombinant Human Keratinocyte Growth Factor (KGF) Source: E.Coli Background: KGF is a potent specific mitogen for many epithelial cells but not for fibroblasts and endothelial cells. KG...