Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Keratinocyte Growth Supplement (HKGS)

Catalog No. S0015
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0015 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0015 Supplier Gibco™ Supplier No. S0015
  • $77.75
    $72.65 / Each of 1

    Save $5.10

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human Keratinocyte Growth Supplement (HKGS)

A sterile, concentrated (100X) solution intended for use with EpiLife™ medium or Medium 154 to culture human epidermal keratinocytes. HKGS is an ionically-balanced supplement containing bovine pituitary extract (BPE), recombinant human insulin-like growth factor-I, hydrocortisone, bovine transferrin, and human epidermal growth factor. Each 5 mL bottle of HKGS contains all of the growth factors, hormones, and tissue extracts necessary for the culture of human epidermal keratinocytes, and is the correct amount of supplement for a 500 mL bottle of medium.

When a 500 mL bottle of medium is supplemented with HKGS, the final concentrations of the components in the supplemented medium are:

  • Bovine pituitary extract (BPE), 0.2% v/v
  • Recombinant human insulin-like growth factor-I, 0.01 μg/mL
  • Hydrocortisone, 0.18μg/mL
  • Bovine transferrin, 5μg/mL
  • Human epidermal growth factor, 0.2 ng/mL
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -20°C).
Cell Type Keratinocyte Cells
Concentration 100 X
Form Liquid
Product Type Growth Supplement
Sterility Sterile
For Use With (Application) Mammalian Cell Culture, Primary Cell Culture
Product Line Cascade Biologics
Quantity 5 mL
Source Human
Species Human
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.