Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Keratinocyte Growth Supplement Kit (HKGS Kit)

Catalog No. S001K
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001K 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001K Supplier Gibco™ Supplier No. S001K
  • $103.00
    $97.65 / Each of 1

    Save $5.35

    5% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human Keratinocyte Growth Supplement Kit (HKGS Kit)

A sterile set of solutions intended for use with EpiLife™ medium or Medium 154 to culture human epidermal keratinocytes. The HKGS Kit provides, in separate vials, all the components of complete HKGS (S-001-5). A vial of gentamycin/amphotericin solution (GA) is also included. Use of GA is optional.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage minus 20 degrees C
Cell Type Keratinocyte Cells
Product Type Growth Supplement
Sterility Sterile
Product Line Cascade Biologics
Quantity 1 Kit
Shipping Condition Dry Ice
Source Human
Species Human
Do I need to add additional supplements to AmnioMAX-II Complete Medium (Cat. No. 11269016)?

No. AmnioMax-II Complete Medium (Cat. No. 1129016) is fully supplemented with antibiotics, L-glutamine, and FBS.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.