Learn More
Biomatik Corporation Human Ki-67 Protein (Ki-67) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10557
Encompass_Preferred
Human Ki-67 Protein (Ki-67) Wide-range ELISA Kit for detecting Ki-67 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P46013. Gene ID: 4288. Target Synonyms: MKI67; KIA; Ki67P; Antigen Identified By Monoclonal Antibody Ki-67. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5815