Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human KIAA0323 (aa 565-668) Control Fragment Recombinant Protein

Catalog No. RP89122
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP89122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP89122 Supplier Invitrogen™ Supplier No. RP89122
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51923. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The KIAA0323 gene, also known as chromosome 18 open reading frame 8, encodes a protein of currently undefined function. Located on chromosome 18 at the 18q21.1 position, KIAA0323 is part of a family of genes that were initially identified through the sequencing of human brain cDNA libraries. Its precise role in cellular processes remains unclear, but like other KIAA proteins, it may be involved in various cellular functions such as signaling pathways, structural organization, and regulatory mechanisms. Further research is required to elucidate the exact biological pathways in which KIAA0323 participates and its potential implications in human health and disease.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O15037
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 23351
Name Human KIAA0323 (aa 565-668) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 6330555F21; 9130227C08Rik; AI605259; KH and NYN domain containing; KH and NYN domain-containing protein; KHNYN; KIAA0323; mKIAA0323; Protein KHNYN
Common Name KIAA0323
Gene Symbol KHNYN
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PFTFVGNLFMVPDDPLGRNGPTLDEFLKKPARTQGSSKAQHPSRGFAEHGKQQQGREEEKGSGGIRKTRETERLRRQLLEVFWGQDHKVDFILQREPYCRDINQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.