Learn More
Amsbio LLC Kidney, cortex Normal FFPE tissue
Catalog No. 502041362
Encompass
Normal human tissue from a clinically-unremarkable donor. Preparation: FFPE block. Tissue is prepared by a certified medical pathologist, fixed in10% neutral formalin for 24 hours and embedded in IHC-grade paraffin. Either surgical or post-mortem (PMI = ...
Catalog No. 50-204-1362 Supplier Amsbio LLC Supplier No. 500026028
Normal human tissue from a clinically-unremarkable donor. Preparation: FFPE block. Tissue is prepared by a certified medical pathologist, fixed in10% neutral formalin for 24 hours and embedded in IHC-grade paraffin. Either surgical or post-mortem (PMI = ...