Learn More
Takara Bio HUMAN KIDNEY RNA 50 UG
SDP
Catalog No. NC9946719
Encompass
Human Kidney Total RNA, 50 microgram, These RNA can be used for a variety of applications, including Atlas Array hybridizations, SMART PCR cDNA synthesis, and RT-PCR analysis
Catalog No. NC9946719 Supplier Takara Bio Supplier No. 636529
Human Kidney Total RNA, 50 microgram, These RNA can be used for a variety of applications, including Atlas Array hybridizations, SMART PCR cDNA synthesis, and RT-PCR analysis