Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human KIFC2 (aa 200-296) Control Fragment Recombinant Protein

Catalog No. RP93887
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP93887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP93887 Supplier Invitrogen™ Supplier No. RP93887
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56098. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Kinesins are a superfamily of microtubule-associated motor proteins involved in a variety of cellular processes including membranous organelle transport and cell division. Kinesin has been found in a variety of organisms and cell types and is subject to spatial and temporal regulation. These proteins have a modular structure which includes a conserved motor domain of approximately 350 amino acids, which is responsible for microtubule binding and ATP hydrolysis. In addition to the motor domain, subfamily members share common domain organization, exhibit sequence similarity, motility properties, and cellular functions outside of the motor domain. An expansive phylogenetic tree of kinesins and kinesin-related proteins (KRPs) has been assembled based on this information. Kinesin C2 (KIFC2) is a C-type kinesin based on the position of the motor domain towards the carboxyl-terminus of the protein and is similar to members of the karyogamy protein 3 (KAR3) family involved in cell division. KIFC2 protein expression has been demonstrated in axons and dendrites of the central and peripheral nervous system. Unlike other C-type motor proteins, KIFC2 appears to play a role in retrograde axonal transport.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96AC6
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 90990
Name Human KIFC2 (aa 200-296) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias KIFC2; kinesin family member C2; kinesin-like protein KIFC2
Common Name KIFC2
Gene Symbol Kifc2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QLILGQLEELKQQLEQQEEELGRLRLGVGATDSEKRVQHLTLENEALKQSLSLMRDLLLHWGPGPPIRAPQEEAEALLELQGRLQEAQDTTEALRAQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.