Learn More
Biomatik Corporation Human Killer Cell Immunoglobulin Like Receptor 3DL1 (KIR3DL1) ELISA Kit
Supplier:  Biomatik Corporation EKU05493
Encompass_Preferred
Human Killer Cell Immunoglobulin Like Receptor 3DL1 (KIR3DL1) ELISA Kit for detecting KIR3DL1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43629. Gene ID: 3811. Target Synonyms: CD158E; HLA-BW4-specific inhibitory NK cell receptor; CD158 antigen-like family member E; Natural killer-associated transcript 3; p70 NK receptor CL-2/CL-11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6708