Learn More
Biomatik Corporation Human Killer Cell Immunoglobulin Like Receptor 2DL3 (KIR2DL3) ELISA Kit
Supplier:  Biomatik Corporation EKU11749
Encompass_Preferred
Human Killer Cell Immunoglobulin Like Receptor 2DL3 (KIR2DL3) ELISA Kit for detecting KIR2DL3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43628. Gene ID: 3804. Target Synonyms: CD158B2; Cl-6; Nkat2; Nkat2a; Nkat2b; p58; CD158 antigen-like family member B2; MHC class I NK cell receptor; Natural killer-associated transcript 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5703