Learn More
Sino Biological Human KIM-1 / TIM1 / HAVCR1 Protein (aa 1-135, His Tag) 1mg
Supplier:  Sino Biological 11051H08H11
Encompass_Preferred
A DNA sequence encoding the N-terminal fragment (Met 1-Val 135) of human KIM1 (AAC39862.1) extracellular domain was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7274